Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026741.2 | NP_081017.1 | 2 | 2071 | ||
GenBank mRNA | AK003337.1 | - | 2 | 664 | ||
AK143550.1 | - | 1 | 3128 | |||
AK148159.1 | - | 1 | 3169 | |||
BC048385.1 | - | 1 | 2401 | 6309639 | ||
BC054068.1 | - | 1 | 2401 |