Gene Symbol: Trnau1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027925.3 | NP_082201.2 | 2 | 262 | ||
GenBank mRNA | AK003534.1 | - | 2 | 240 | ||
AK152986.1 | - | 2 | 211 | |||
AK171407.1 | - | 1 | 803 | |||
BC005795.1 | - | 2 | 219 | |||
BC048840.1 | - | 2 | 213 | 5008141 | ||
BC055454.1 | - | 2 | 225 | 4014366 | ||
BY283216.1 | - | 2 | 263 | |||
CJ166630.1 | - | 2 | 225 |