Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172549.3 | NP_766137.2 | 20, 21 | 3377 | ||
GenBank mRNA | BC046407.1 | - | 3, 4 | 459 | ||
BC057551.1 | - | 20, 21 | 3305 | |||
BC150825.1 | - | 19, 20 | 2941 | 9087736 |