Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025642.1 | NP_079918.1 | 5 | 1164 | ||
GenBank mRNA | AK011719.1 | - | 5 | 1014 | ||
AK012533.1 | - | 5 | 1165 | |||
AK014084.1 | - | 5 | 1134 | |||
AK163310.1 | - | 5 | 1198 | |||
BC079900.1 | - | 5 | 1176 | 30602551 |