Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020406.3 | NP_065139.2 | 4, 5 | 566 | ||
XM_017027021.1 | XP_016882510.1 | 4, 5 | 550 | |||
XM_017027022.1 | XP_016882511.1 | 4, 5 | 550 | |||
GenBank mRNA | AB237911.1 | - | 4, 5 | 518 | ||
AF146747.1 | - | 4, 5 | 545 | |||
AJ290452.1 | - | 4, 5 | 537 | |||
AJ305326.1 | - | 4, 5 | 656 | |||
AJ310433.1 | - | 4, 5 | 537 | |||
AK225226.1 | - | 4, 5 | 539 | |||
AK290965.1 | - | 4, 5 | 541 | |||
AY358932.1 | - | 4, 5 | 526 | |||
BC029167.1 | - | 4, 5 | 528 | 5182826 | ||
BT020111.1 | - | 4, 5 | 510 | |||
CD514938.1 | - | 4, 5 | 514 | |||
DA728784.1 | - | 4, 5 | 566 |