Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sstr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191008.1 | NP_001177937.1 | 2 | 1745 | ||
NM_011425.3 | NP_035555.1 | 3 | 1897 | |||
GenBank mRNA | AK133609.1 | - | 3 | 1863 | ||
AK133767.1 | - | 2 | 1673 | |||
BC150805.1 | - | 2 | 1729 | 9087716 | ||
GQ359776.1 | - | 2 | 540 |
Set Membership: |
G-protein coupled receptor |