Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013376.2 | NP_001013394.2 | 2 | 172 | ||
GenBank mRNA | AK049133.1 | - | 2 | 117 | ||
AK087782.1 | - | 2 | 173 | |||
AK139023.1 | - | 2 | 100 | |||
BC051448.1 | - | 2 | 147 | 30060460 |