Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013376.2 | NP_001013394.2 | 2 | 931 | ||
GenBank mRNA | AK139023.1 | - | 2 | 859 | ||
BC051448.1 | - | 2 | 906 | 30060460 |