Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033299.3 | NP_001028471.1 | 5 | 3635 | ||
NM_001159683.1 | NP_001153155.1 | 6 | 3620 | |||
GenBank mRNA | AK030249.1 | - | 4 | 2092 | ||
AK139547.1 | - | 6 | 3622 | |||
AK156475.1 | - | 5 | 3636 | |||
AK163381.1 | - | 6 | 3627 | |||
BC046393.1 | - | 2 | 629 | |||
BC116329.1 | - | 4 | 3114 | 40096805 | ||
BC116330.1 | - | 4 | 3114 | 40096806 |