Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033299.3 | NP_001028471.1 | 6 | 3886 | ||
NM_001159683.1 | NP_001153155.1 | 7 | 3871 | |||
GenBank mRNA | AK139547.1 | - | 7 | 3873 | ||
AK156475.1 | - | 6 | 3887 | |||
BC046393.1 | - | 3 | 880 |