Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033321.1 | NP_001028493.1 | 7 | 1540 | ||
GenBank mRNA | AK030046.1 | - | 8 | 1629 | ||
AK155425.1 | - | 7 | 1540 | |||
BC100402.1 | - | 7 | 1524 | 30106220 |