Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tatdn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033463.3 | NP_001028635.2 | 4 | 1189 | ||
GenBank mRNA | AK146408.1 | - | 3 | 1121 | ||
AK156197.1 | - | 4 | 1190 | |||
AK220217.1 | - | 3 | 1303 | |||
BC141065.1 | - | 4 | 1307 | 9055677 | ||
BC145318.1 | - | 4 | 1307 | 9053873 |