Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012725.2 | NP_001012743.2 | 3 | 428 | ||
GenBank mRNA | AY541526.1 | - | 3 | 428 | ||
BC141216.1 | - | 3 | 406 | 9055828 |