Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trcg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014398.2 | NP_001014420.2 | 2 | 176 | ||
GenBank mRNA | AY701870.1 | - | 2 | 176 |