Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-T3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY989874.1 | - | 4 | 680 | ||
AY989875.1 | - | 4 | 663 | |||
AY989876.1 | - | 2 | 134 | |||
AY989877.1 | - | 2 | 149 |