Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020314.5 | NP_064710.4 | 25 | 2394 | ||
XM_006721065.3 | XP_006721128.1 | 23 | 2145 | |||
XM_011545896.2 | XP_011544198.1 | 25 | 2095 | |||
XM_011545897.2 | XP_011544199.1 | 18 | 1462 | |||
GenBank mRNA | AF461052.2 | - | 19 | 1890 | ||
AK024693.1 | - | 21 | 1664 | |||
AK290286.1 | - | 25 | 2091 | |||
AL136744.1 | - | 12 | 1007 | |||
AL832159.1 | - | 25 | 2090 | |||
BC014900.2 | - | 24 | 2062 | 3909068 | ||
BC050464.1 | - | 25 | 2134 | 6061139 | ||
BC058845.1 | - | 25 | 2093 | 6452778 | ||
BX537867.1 | - | 25 | 2106 |