Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027830.2 | NP_082106.2 | 5 | 1128 | ||
GenBank mRNA | AK018494.1 | - | 4 | 968 | ||
AK046636.1 | - | 4 | 912 | |||
AK139112.1 | - | 4 | 912 |