Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028599.2 | NP_082875.1 | 17 | 1935 | ||
GenBank mRNA | AK010772.1 | - | 17 | 1935 | ||
AK152412.1 | - | 17 | 1936 | |||
AK163221.1 | - | 17 | 1909 | |||
AK168032.1 | - | 17 | 1929 | |||
AK168163.1 | - | 17 | 1937 | |||
AK169932.1 | - | 17 | 1933 | |||
BC038920.1 | - | 3 | 252 | |||
BC049925.1 | - | 5 | 1393 | |||
BC055483.1 | - | 6 | 836 | |||
BC085177.1 | - | 17 | 1965 | |||
BC103800.1 | - | 3 | 268 | |||
BC117880.2 | - | 17 | 1965 | 40096010 | ||
BC117881.1 | - | 17 | 1962 | 40096014 |