Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287214.1 | NP_001274143.1 | 8 | 1615 | ||
NM_001287215.1 | NP_001274144.1 | 9 | 1465 | |||
NM_001287216.1 | NP_001274145.1 | 5 | 1342 | |||
NM_001634.5 | NP_001625.2 | 9 | 1702 | |||
NR_109768.1 | 6 | 1385 | ||||
GenBank mRNA | AJ420527.1 | - | 6 | 1360 | ||
AJ420567.1 | - | 1 | 201 | |||
AK125644.1 | - | 4 | 2615 | |||
AK130474.1 | - | 1 | 515 | |||
AL832698.1 | - | 6 | 1384 | |||
BC000171.2 | - | 9 | 1475 | 2900587 | ||
BX640599.1 | - | 9 | 1702 | |||
M21154.1 | - | 9 | 1628 |