Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012631.1 | NP_001012649.1 | 3, 4 | 301 | ||
NM_001012632.1 | NP_001012650.1 | 2, 3 | 100 | |||
NM_001012633.1 | NP_001012651.1 | 2, 3 | 234 | |||
NM_001012634.1 | NP_001012652.1 | 2, 3 | 101 | |||
NM_001012635.1 | NP_001012653.1 | 2, 3 | 96 | |||
NM_001012636.1 | NP_001012654.1 | 1, 2 | 51 | |||
NM_001012718.1 | NP_001012736.1 | 2, 3 | 237 | |||
NM_001308078.1 | NP_001295007.1 | 2, 3 | 97 | |||
NM_004221.4 | NP_004212.4 | 2, 3 | 234 | |||
GenBank mRNA | AK296143.1 | - | 2, 3 | 106 | ||
AK307158.1 | - | 1, 2 | 480 | |||
AK310053.1 | - | 2, 3 | 100 | |||
AY495331.1 | - | 2, 3 | 224 | |||
AY495332.1 | - | 2, 3 | 224 | |||
BC009401.2 | - | 2, 3 | 234 | 4300407 | ||
BC018782.1 | - | 2, 3 | 81 | 4873608 | ||
BC105602.1 | - | 2, 3 | 102 | 6496101 | ||
BF568456.1 | - | 2, 3 | 235 | |||
BG748458.1 | - | 2, 3 | 180 | |||
BM822755.1 | - | 3, 4 | 209 | |||
EU339107.1 | - | 1, 2 | 23 | |||
FJ985780.1 | - | 2, 3 | 188 | |||
JN546102.1 | - | 1, 2 | 23 | |||
M59807.1 | - | 2, 3 | 82 |