Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFITM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021034.2 | NP_066362.2 | 2 | 562 | ||
NR_049759.1 | 2 | 557 | ||||
GenBank mRNA | AK292173.1 | - | 2 | 506 | ||
BC006794.1 | - | 2 | 520 | 2986145 | ||
BC008417.1 | - | 2 | 505 | 4075453 | ||
BC022439.1 | - | 2 | 511 | 4282809 | ||
BC070243.1 | - | 2 | 496 | 30401041 | ||
BQ937265.1 | - | 2 | 365 | |||
X57352.1 | - | 2 | 698 |