Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014322.2 | NP_055137.2 | 1 | 440 | ||
GenBank mRNA | AF140242.1 | - | 1 | 350 | ||
AF303588.1 | - | 1 | 397 | |||
BC036773.1 | - | 1 | 392 | 5723543 | ||
BC051900.1 | - | 1 | 314 | |||
BC062622.1 | - | 1 | 381 | |||
BC073872.1 | - | 1 | 365 |
Set Membership: |
G-protein coupled receptor |