Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FAM178B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122646.2 | NP_001116118.2 | 16 | 2217 | ||
NM_001172667.1 | NP_001166138.1 | 4 | 418 | |||
NM_016490.4 | NP_057574.2 | 4 | 536 | |||
XM_011511294.2 | XP_011509596.1 | 15 | 2145 | |||
XM_017004263.1 | XP_016859752.1 | 17 | 2382 | |||
XM_017004264.1 | XP_016859753.1 | 17 | 2343 | |||
XM_017004266.1 | XP_016859755.1 | 7 | 790 | |||
XM_017004267.1 | XP_016859756.1 | 4 | 395 | |||
XM_017004268.1 | XP_016859757.1 | 4 | 421 | |||
GenBank mRNA | AK122705.1 | - | 10 | 1096 | ||
AK308698.1 | - | 10 | 1099 | |||
BC038583.1 | - | 4 | 497 | |||
BC039488.1 | - | 14 | 1484 | |||
BC051379.1 | - | 4 | 536 | |||
BX103404.1 | - | 4 | 396 |