Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301769.1 | NP_001288698.1 | 6 | 1369 | ||
NM_001301771.1 | NP_001288700.1 | 5 | 1338 | |||
NM_001301773.1 | NP_001288702.1 | 4 | 1306 | |||
NM_001301774.1 | NP_001288703.1 | 4 | 1199 | |||
NM_001301775.1 | NP_001288704.1 | 6 | 1410 | |||
NM_020422.5 | NP_065155.3 | 5 | 1445 | |||
NR_125971.1 | 7 | 1546 | ||||
XM_005255439.4 | XP_005255496.1 | 6 | 1097 | |||
XM_006721066.2 | XP_006721129.1 | 6 | 1545 | |||
XM_006721069.3 | XP_006721132.1 | 5 | 1406 | |||
GenBank mRNA | AF070596.1 | - | 7 | 1546 | ||
AK074667.1 | - | 5 | 1143 | |||
AK293557.1 | - | 6 | 1098 | |||
BC015812.1 | - | 4 | 982 | 3920903 |