Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM109B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002034.2 | NP_001002034.2 | 3 | 2307 | ||
XM_005261373.3 | XP_005261430.1 | 3 | 2445 | |||
GenBank mRNA | BC029776.1 | - | 1 | 1967 | ||
BX648402.1 | - | 3 | 2313 |