Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318031.1 | NP_001304960.1 | 2 | 297 | ||
NM_020436.4 | NP_065169.1 | 2 | 297 | |||
XM_005260467.3 | XP_005260524.1 | 2 | 278 | |||
XM_011528921.2 | XP_011527223.1 | 4 | 473 | |||
XM_011528922.2 | XP_011527224.1 | 3 | 432 | |||
GenBank mRNA | AK001666.1 | - | 2 | 249 | ||
AY170621.1 | - | 2 | 182 | |||
AY172738.1 | - | 2 | 182 | |||
BC111714.1 | - | 2 | 238 | 40008661 | ||
DA666635.1 | - | 2 | 300 | |||
DB066881.1 | - | 2 | 283 |