Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001370.1 | NP_001361.1 | 48 | 7944 | ||
XM_006711956.2 | XP_006712019.1 | 48 | 8049 | |||
XM_011532649.2 | XP_011530951.1 | 48 | 7898 | |||
XM_011532650.2 | XP_011530952.1 | 48 | 7899 | |||
XM_011532652.1 | XP_011530954.1 | 47 | 7818 | |||
XM_011532653.1 | XP_011530955.1 | 47 | 7797 | |||
XM_011532654.1 | XP_011530956.1 | 47 | 7677 | |||
XM_011532655.2 | XP_011530957.1 | 44 | 6966 | |||
XM_011532657.1 | XP_011530959.1 | 42 | 6705 | |||
XM_017003521.1 | XP_016859010.1 | 46 | 7671 | |||
XM_017003522.1 | XP_016859011.1 | 43 | 7770 | |||
XM_017003523.1 | XP_016859012.1 | 43 | 6937 | |||
GenBank mRNA | AB051484.2 | - | 12 | 1879 | ||
AB290165.1 | - | 14 | 2083 | |||
BC117259.1 | - | 1 | 76 |