Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB16-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_049729.1 | 4 | 1517 | |||
GenBank mRNA | AK055685.1 | - | 4 | 1471 | ||
BC009448.1 | - | 1 | 599 | 3530891 | ||
BC100964.2 | - | 1 | 923 | 40004836 | ||
BC100965.1 | - | 1 | 922 | 40004837 | ||
BC100966.1 | - | 1 | 923 | 40004838 | ||
BC100967.1 | - | 1 | 922 | 40004839 |