Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079684.2 | NP_001073152.1 | 4 | 588 | ||
NM_001079685.2 | NP_001073153.1 | 4 | 567 | |||
NM_001287335.1 | NP_001274264.1 | 2 | 301 | |||
NM_001287336.1 | NP_001274265.1 | 3 | 480 | |||
NM_020444.4 | NP_065177.2 | 5 | 675 | |||
XM_005265941.1 | XP_005265998.1 | 4 | 488 | |||
XM_005265946.4 | XP_005266003.1 | 4 | 583 | |||
XM_011534595.1 | XP_011532897.1 | 4 | 450 | |||
XM_011534596.1 | XP_011532898.1 | 3 | 399 | |||
XM_017009651.1 | XP_016865140.1 | 3 | 384 | |||
GenBank mRNA | AK315428.1 | - | 5 | 632 | ||
AL390217.1 | - | 4 | 546 | |||
BC010448.1 | - | 5 | 610 | 4155679 | ||
BC017097.1 | - | 5 | 632 | 3529175 | ||
BX648859.1 | - | 5 | 674 | |||
CR457331.1 | - | 3 | 302 | |||
DA789543.1 | - | 4 | 557 | |||
DQ073392.1 | - | 3 | 302 | |||
HY114499.1 | - | 2 | 259 | |||
HY119081.1 | - | 3 | 480 |