Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145053.1 | NP_001138525.1 | 7 | 1094 | ||
NM_198529.3 | NP_940931.2 | 7 | 1215 | |||
NR_026738.1 | 7 | 1215 | ||||
XM_011524759.1 | XP_011523061.1 | 7 | 1221 | |||
XM_011524763.2 | XP_011523065.1 | 7 | 1221 | |||
XM_011524765.1 | XP_011523067.1 | 7 | 1221 | |||
XM_011524766.2 | XP_011523068.1 | 2 | 161 | |||
XM_011524768.2 | XP_011523070.1 | 7 | 1221 | |||
XM_017024591.1 | XP_016880080.1 | 4 | 300 | |||
XM_017024592.1 | XP_016880081.1 | 7 | 1215 | |||
GenBank mRNA | AK128126.1 | - | 4 | 441 | ||
AK299608.1 | - | 7 | 1047 | |||
AK302745.1 | - | 6 | 524 | |||
AK303060.1 | - | 6 | 429 | |||
BC110608.1 | - | 4 | 418 | |||
BC110609.1 | - | 4 | 418 | |||
BC119807.1 | - | 4 | 418 | |||
BC137519.1 | - | 7 | 1051 | 9021519 | ||
BX648689.1 | - | 7 | 1153 | |||
DB076773.1 | - | 4 | 441 |