Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030311.2 | NP_001025482.1 | 8 | 1243 | ||
NM_001030312.2 | NP_001025483.1 | 4 | 826 | |||
NM_001030313.2 | NP_001025484.1 | 5 | 958 | |||
NM_001160277.1 | NP_001153749.1 | 7 | 1111 | |||
NM_201548.4 | NP_963842.1 | 7 | 1165 | |||
NR_027689.1 | 5 | 969 | ||||
NR_027690.1 | 6 | 1101 | ||||
GenBank mRNA | AJ640141.1 | - | 7 | 1082 | ||
AJ697855.1 | - | 7 | 1064 | |||
AJ697856.1 | - | 6 | 1000 | |||
AJ697858.1 | - | 5 | 884 | |||
AK129976.1 | - | 4 | 408 | |||
AK293844.1 | - | 7 | 1112 | |||
AY357073.1 | - | 7 | 1064 | |||
AY690329.1 | - | 8 | 1142 | |||
AY690330.1 | - | 4 | 725 | |||
AY690331.1 | - | 5 | 857 | |||
AY690332.1 | - | 5 | 868 | |||
AY690333.1 | - | 6 | 1000 | |||
BC137498.1 | - | 7 | 1091 | 9021498 | ||
BC137499.1 | - | 7 | 1091 | 9021499 | ||
HQ426666.1 | - | 7 | 971 | |||
HQ426667.1 | - | 6 | 907 | |||
HQ426668.1 | - | 5 | 775 | |||
HQ426669.1 | - | 5 | 764 | |||
HQ426670.1 | - | 4 | 632 | |||
HQ426671.1 | - | 8 | 971 | |||
HQ426672.1 | - | 7 | 893 | |||
HQ426673.1 | - | 6 | 829 | |||
HQ426674.1 | - | 5 | 697 | |||
HQ426675.1 | - | 5 | 686 | |||
HQ426676.1 | - | 4 | 554 | |||
HQ426677.1 | - | 5 | 501 | |||
HQ426678.1 | - | 8 | 1218 | |||
HQ426679.1 | - | 6 | 826 | |||
JF290420.1 | - | 7 | 1051 |