Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYSRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199001.3 | NP_945352.3 | 2 | 645 | ||
GenBank mRNA | BC018787.1 | - | 2 | 645 | ||
BC052297.1 | - | 2 | 88 | 6293780 | ||
BE563091.1 | - | 1 | 56 |