Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300861.1 | NP_001287790.1 | 2 | 415 | ||
NM_020470.2 | NP_065203.2 | 2 | 415 | |||
XM_005273720.3 | XP_005273777.1 | 2 | 391 | |||
XM_017017139.1 | XP_016872628.1 | 2 | 391 | |||
XM_017017140.1 | XP_016872629.1 | 2 | 391 | |||
XM_017017141.1 | XP_016872630.1 | 2 | 391 | |||
GenBank mRNA | AF004876.1 | - | 2 | 348 | ||
AK226003.1 | - | 2 | 393 | |||
AK291124.1 | - | 2 | 393 | |||
AK308988.1 | - | 2 | 385 | |||
AK310521.1 | - | 2 | 294 | |||
BC001299.1 | - | 2 | 366 | 3451489 | ||
BG397210.1 | - | 2 | 322 | |||
CR456710.1 | - | 2 | 233 | |||
DB085949.1 | - | 2 | 415 | |||
S83365.1 | - | 1 | 206 |