Search Thermo Fisher Scientific
Gene Symbol: FGF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020638.2 | NP_065689.1 | 2 | 413 | ||
GenBank mRNA | AB037973.1 | - | 2 | 267 | ||
AB047858.1 | - | 2 | 399 | |||
AF263537.1 | - | 2 | 413 | |||
AY358323.1 | - | 2 | 300 | |||
BC069333.1 | - | 2 | 341 | 7262193 | ||
BC096713.1 | - | 2 | 306 | 40000232 | ||
BC098147.1 | - | 2 | 306 | 40000231 | ||
BC098252.1 | - | 2 | 306 | 40000230 |