Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020650.2 | NP_065701.2 | 2 | 653 | ||
XM_005259089.3 | XP_005259146.1 | 3 | 842 | |||
XM_011527143.1 | XP_011525445.1 | 3 | 842 | |||
XM_017027023.1 | XP_016882512.1 | 2 | 927 | |||
GenBank mRNA | AF183423.1 | - | 1 | 238 | ||
AY195859.1 | - | 2 | 653 | |||
AY358423.1 | - | 2 | 240 | |||
BC013436.1 | - | 2 | 236 | 3889718 |