Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077203.2 | NP_001070671.1 | 3 | 239 | ||
NM_001282801.1 | NP_001269730.1 | 3 | 239 | |||
NM_001282802.1 | NP_001269731.1 | 2 | 189 | |||
NM_001282803.1 | NP_001269732.1 | 2 | 189 | |||
NM_001282804.1 | NP_001269733.1 | 2 | 189 | |||
NM_020654.4 | NP_065705.3 | 3 | 239 | |||
XM_005247662.4 | XP_005247719.1 | 3 | 225 | |||
XM_011513038.2 | XP_011511340.1 | 3 | 225 | |||
XM_011513039.2 | XP_011511341.1 | 2 | 150 | |||
XM_011513040.2 | XP_011511342.1 | 3 | 225 | |||
XM_011513041.2 | XP_011511343.1 | 3 | 225 | |||
XM_011513042.2 | XP_011511344.1 | 3 | 225 | |||
XM_011513045.2 | XP_011511347.1 | 3 | 226 | |||
XM_017006924.1 | XP_016862413.1 | 3 | 113 | |||
XM_017006925.1 | XP_016862414.1 | 3 | 204 | |||
XM_017006926.1 | XP_016862415.1 | 2 | 137 | |||
XM_017006927.1 | XP_016862416.1 | 2 | 137 | |||
XM_017006928.1 | XP_016862417.1 | 3 | 226 | |||
XM_017006929.1 | XP_016862418.1 | 3 | 228 | |||
GenBank mRNA | AF199458.1 | - | 2 | 187 | ||
AK297356.1 | - | 150 | ||||
AL136599.1 | - | 3 | 265 | |||
AL833577.1 | - | 3 | 205 | |||
BC062674.1 | - | 3 | 186 | |||
BC129988.1 | - | 3 | 166 | 40128878 | ||
BX537943.1 | - | 2 | 133 |