Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASKIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020764.3 | NP_065815.1 | 3, 4 | 274 | ||
GenBank mRNA | AF451977.1 | - | 3, 4 | 340 |