Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018070.2 | NP_001018080.1 | 12 | 1891 | ||
NM_020441.2 | NP_065174.1 | 11 | 1869 | |||
GenBank mRNA | AK000860.1 | - | 11 | 1853 | ||
AL162072.1 | - | 6 | 1142 | |||
BC006449.1 | - | 11 | 1819 | 3958930 |