Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322854.1 | NP_001309783.1 | 8, 9 | 1321 | ||
NM_001322855.1 | NP_001309784.1 | 9, 10 | 1463 | |||
NM_001322856.1 | NP_001309785.1 | 9, 10 | 1420 | |||
NM_001322857.1 | NP_001309786.1 | 8, 9 | 1241 | |||
NM_001322858.1 | NP_001309787.1 | 9, 10 | 1338 | |||
NM_001322859.1 | NP_001309788.1 | 6, 7 | 947 | |||
NM_001322860.1 | NP_001309789.1 | 5, 6 | 887 | |||
NM_001322861.1 | NP_001309790.1 | 5, 6 | 597 | |||
NM_001322862.1 | NP_001309791.1 | 9, 10 | 1298 | |||
NM_001322863.1 | NP_001309792.1 | 8, 9 | 1270 | |||
NM_001322864.1 | NP_001309793.1 | 7, 8 | 838 | |||
NM_001322866.1 | NP_001309795.1 | 8, 9 | 1161 | |||
NM_020448.4 | NP_065181.1 | 8, 9 | 1183 | |||
NR_136506.1 | 7, 8 | 1091 | ||||
XM_011541808.2 | XP_011540110.1 | 7, 8 | 1098 | |||
XM_017001868.1 | XP_016857357.1 | 7, 8 | 833 | |||
XM_017001869.1 | XP_016857358.1 | 8, 9 | 1168 | |||
XM_017001870.1 | XP_016857359.1 | 9, 10 | 1433 | |||
GenBank mRNA | AK312433.1 | - | 8, 9 | 1137 | ||
BC063583.1 | - | 8, 9 | 1113 | 4386115 | ||
BX640883.1 | - | 9, 10 | 1245 | |||
U90918.1 | - | 8, 9 | 1074 |