Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRVI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098579.2 | NP_001092049.2 | 2 | 361 | ||
NM_001100163.2 | NP_001093633.1 | 3 | 642 | |||
NM_001206880.1 | NP_001193809.1 | 3 | 695 | |||
NM_001206881.1 | NP_001193810.1 | 2 | 537 | |||
NM_130385.3 | NP_569056.4 | 3 | 695 | |||
GenBank mRNA | AF081249.1 | - | 2 | 434 | ||
AF081250.1 | - | 2 | 162 | |||
AK296336.1 | - | 3 | 459 | |||
AK300358.1 | - | 2 | 346 | |||
AK301296.1 | - | 3 | 306 | |||
AK304702.1 | - | 3 | 329 | |||
BC117127.1 | - | 3 | 232 | 40125678 | ||
DA127310.1 | - | 2 | 514 | |||
DA797538.1 | - | 2 | 361 | |||
DB132030.1 | - | 2 | 154 | |||
DB167290.1 | - | 4 | 444 | |||
DB445355.2 | - | 4 | 578 |