Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000979.1 | NP_001000979.1 | 1 | 408 | ||
GenBank mRNA | AF091577.1 | - | 1 | 219 |