Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC298116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003409.1 | NP_001003409.1 | 6 | 585 | ||
GenBank mRNA | M26836.1 | - | 6 | 585 |
Gene Symbol: LOC259246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M26835.1 | - | 6 | 600 |