Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Esrrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008516.2 | NP_001008516.2 | 5 | 965 | ||
XM_006240374.2 | XP_006240436.1 | 6 | 1427 | |||
XM_006240375.2 | XP_006240437.1 | 6 | 1428 | |||
XM_008764759.1 | XP_008762981.1 | 6 | 2663 | |||
GenBank mRNA | AY383731.1 | - | 5 | 965 |