Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip5k1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009967.2 | NP_001009967.2 | 5 | 456 | ||
NM_001033970.1 | NP_001029142.1 | 5 | 456 | |||
XM_006240935.2 | XP_006240997.1 | 5 | 499 | |||
XM_008765101.1 | XP_008763323.1 | 5 | 499 | |||
XM_008765102.1 | XP_008763324.1 | 5 | 499 | |||
XM_008765103.1 | XP_008763325.1 | 5 | 499 | |||
GenBank mRNA | AY850259.1 | - | 5 | 395 | ||
BC101909.1 | - | 5 | 428 | 7933923 | ||
CB557931.1 | - | 5 | 467 | |||
CK366332.1 | - | 5 | 445 |