Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014028.1 | NP_001014050.1 | 2 | 316 | ||
XM_006231162.2 | XP_006231224.1 | 2 | 406 | |||
GenBank mRNA | BC079229.1 | - | 2 | 316 | 7132456 |