Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNNT3 SIRNA Assays ›
Gene Symbol:
Tnnt3
Gene Name:
troponin T type 3 (skeletal, fast)
Gene Aliases:
Tnt
Chromosome Location:
Chr.1: 215666628 - 215683628 on Build Rnor_6.0
UniGene ID:
Rn.15488
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
286796
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tnnt3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270664.1
NP_001257593.1
9
358
NM_001270665.1
NP_001257594.1
11
391
NM_001270666.1
NP_001257595.1
11
391
NM_001270668.1
NP_001257597.1
9
358
NM_001270670.1
NP_001257599.1
10
373
NM_001270671.1
NP_001257600.1
10
373
NM_001270672.1
NP_001257601.1
9
364
NM_001270673.1
NP_001257602.1
10
376
NM_001270675.1
NP_001257604.1
9
364
NM_001270676.1
NP_001257605.1
10
376
NM_001270677.1
NP_001257606.1
8
346
NM_001270678.1
NP_001257607.1
10
379
NM_031532.2
NP_113720.1
10
379
XM_006230681.2
XP_006230743.1
9
321
XM_006230683.2
XP_006230745.1
8
309
XM_006230685.2
XP_006230747.1
7
291
XM_006230686.2
XP_006230748.1
7
291
XM_008760080.1
XP_008758302.1
12
425
XM_008760081.1
XP_008758303.1
12
425
XM_008760082.1
XP_008758304.1
11
375
XM_008760083.1
XP_008758305.1
11
372
XM_008760084.1
XP_008758306.1
10
398
XM_008760085.1
XP_008758307.1
10
363
XM_008760086.1
XP_008758308.1
9
345
GenBank mRNA
CB612473.1
-
11
340
DQ062204.1
-
10
275
DQ273678.1
-
10
275
FQ214650.1
-
10
339
FQ214722.1
-
9
326
FQ214724.1
-
10
340
FQ214766.1
-
9
320
FQ214833.1
-
10
341
FQ215030.1
-
9
320
FQ215059.1
-
9
351
FQ215106.1
-
9
320
FQ215114.1
-
9
326
FQ215252.1
-
10
338
FQ215316.1
-
9
340
FQ215327.1
-
11
353
FQ215374.1
-
10
366
FQ215598.1
-
11
350
FQ215602.1
-
9
353
FQ215694.1
-
9
358
FQ215723.1
-
11
392
FQ215881.1
-
11
353
FQ215885.1
-
11
341
FQ215897.1
-
10
366
FQ215955.1
-
9
320
FQ216024.1
-
6
320
FQ216051.1
-
9
320
FQ216059.1
-
11
348
FQ216097.1
-
10
334
FQ216131.1
-
9
351
FQ216208.1
-
10
338
FQ216324.1
-
9
320
FQ216367.1
-
9
320
FQ216417.1
-
10
339
FQ216467.1
-
9
320
FQ216480.1
-
9
326
FQ216535.1
-
10
335
FQ216561.1
-
6
320
FQ216694.1
-
10
335
FQ216711.1
-
9
357
FQ216750.1
-
6
211
FQ216763.1
-
7
311
FQ216784.1
-
9
326
FQ216866.1
-
11
353
FQ216925.1
-
1
82
FQ217147.1
-
9
319
FQ217846.1
-
11
385
FQ217900.1
-
10
366
FQ223840.1
-
9
320
FQ223999.1
-
9
320
FQ224006.1
-
9
322
FQ224345.1
-
1
51
FQ224794.1
-
9
320
FQ224836.1
-
9
320
FQ224839.1
-
10
338
FQ224892.1
-
10
341
FQ224966.1
-
10
324
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal muscle contraction
muscle contraction
regulation of muscle contraction
regulation of striated muscle contraction
metabolic process
regulation of ATPase activity
actin binding
tropomyosin binding
troponin C binding
calcium-dependent ATPase activity
troponin I binding
troponin T binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0