Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175601.1 | NP_783191.1 | 1 | 799 | ||
XM_006227718.2 | XP_006227780.2 | 2 | 865 | |||
XM_006227719.2 | XP_006227781.1 | 2 | 1287 | |||
XM_008758615.1 | XP_008756837.1 | 3 | 1445 |
Set Membership: |
![]() |