Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001805.2 | NP_001001805.2 | 1 | 371 | ||
GenBank mRNA | BC106977.2 | - | 1 | 371 | 40042102 |