Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm13040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113736.1 | NP_001107208.1 | 3 | 477 |
Gene Symbol: BC080695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007579.3 | NP_001007580.1 | 477 | |||
GenBank mRNA | BC080695.1 | - | 478 | 30458858 |
Gene Symbol: Gm13043 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039595.2 | NP_001034684.1 | 2 | 430 | ||
GenBank mRNA | BC099534.1 | - | 2 | 430 | 30462179 |
Gene Symbol: Gm13057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113735.1 | NP_001107207.1 | 477 |
Gene Symbol: Pramef25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126315.2 | NP_001119787.1 | 2 | 452 |