Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar7e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010835.1 | NP_001010835.1 | 1 | 287 | ||
GenBank mRNA | BC139110.1 | - | 1 | 353 | 8862132 | |
BC145816.1 | - | 1 | 353 | 40131119 |
Set Membership: |
![]() |